Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
384061
Gene Name
fibronectin type III domain containing 5
Gene Aliases
1500001L03Rik, AI836596, C87088, PeP, Pxp
Chromosome Location
Chr. 4: 129136922 - 129144593 on Build GRCm38
UniGene ID
Mm.44075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027402.3 NP_081678.1 6 2570
GenBank mRNA AJ401376.1 - 6 2571
AK137327.1 - 1 1256

View Full Product Details