Gene Transcripts

Gene Symbol
Cnep1r1
Entrez Gene ID
382030
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
5033428A16Rik, NEP1-R1, Tmem188
Chromosome Location
Chr. 8: 88118735 - 88135197 on Build GRCm38
UniGene ID
Mm.216574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029074.3 NP_083350.2 6 837
GenBank mRNA AK017198.1 - 5 1340
BC018498.1 - 5 735

View Full Product Details