Gene Transcripts
- Gene Symbol
- Sept5
- Entrez Gene ID
- 18951
- Gene Name
- septin 5
- Gene Aliases
- Cdcrel-1, Cdcrel1, Pnutl1
- Chromosome Location
- Chr. 16: 18621811 - 18629938 on Build GRCm38
- UniGene ID
- Mm.20365
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_213614.2 |
NP_998779.2
|
7, 8 |
705 |
|
GenBank mRNA |
AF033350.1 |
- |
5, 6 |
552 |
|
AK134700.1 |
- |
7, 8 |
642 |
|
BC059848.1 |
- |
5, 6 |
583 |
|
BC079631.1 |
- |
1, 2 |
19 |
|
BC094347.1 |
- |
7, 8 |
678 |
|
BC141073.1 |
- |
7, 8 |
715 |
9055685 |
BC145331.1 |
- |
7, 8 |
715 |
9053887 |
BC145332.1 |
- |
7, 8 |
728 |
9053888 |
CA945297.1 |
- |
7, 8 |
721 |
|
View Full Product Details