Gene Transcripts

Gene Symbol
Fam19a3
Entrez Gene ID
329731
Gene Name
family with sequence similarity 19, member A3
Gene Aliases
7530404M11Rik, TAFA3, Tafa-3
Chromosome Location
Chr. 3: 104767405 - 104777547 on Build GRCm38
UniGene ID
Mm.389494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183224.3 NP_899047.1 7 3679
GenBank mRNA AK078681.1 - 7 3682

View Full Product Details