Gene Transcripts
- Gene Symbol
- NCAPH
- Entrez Gene ID
- 23397
- Gene Name
- non-SMC condensin I complex subunit H
- Gene Aliases
- BRRN1, CAP-H
- Chromosome Location
- Chr. 2: 96335741 - 96377095 on Build GRCh38
- UniGene ID
- Hs.308045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281710.1 |
NP_001268639.1
|
5, 6 |
647 |
|
NM_001281711.1 |
NP_001268640.1
|
5, 6 |
608 |
|
NM_001281712.1 |
NP_001268641.1
|
4, 5 |
427 |
|
NM_015341.4 |
NP_056156.2
|
5, 6 |
680 |
|
XM_005263908.3 |
XP_005263965.1
|
5, 6 |
620 |
|
XM_006712388.3 |
XP_006712451.1
|
5, 6 |
620 |
|
GenBank mRNA |
AK291053.1 |
- |
5, 6 |
621 |
|
AK299248.1 |
- |
5, 6 |
549 |
|
AK303725.1 |
- |
4, 5 |
372 |
|
AK308050.1 |
- |
5, 6 |
587 |
|
AK310649.1 |
- |
5, 6 |
548 |
|
BC024211.2 |
- |
5, 6 |
634 |
3504782 |
BI559873.1 |
- |
5, 6 |
668 |
|
D38553.1 |
- |
5, 6 |
610 |
|
View Full Product Details