Gene Transcripts

Gene Symbol
ANGEL1
Entrez Gene ID
23357
Gene Name
angel homolog 1
Gene Aliases
Ccr4e, KIAA0759
Chromosome Location
Chr. 14: 76786473 - 76812947 on Build GRCh38
UniGene ID
Hs.497448
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015305.3 NP_056120.2 10 2557
GenBank mRNA AB018302.1 - 10 2454
AK025609.1 - 1 483
AK301069.1 - 3 949
AL137268.1 - 10 2455
BC028714.2 - 10 2504 4830897

View Full Product Details