Gene Transcripts

Gene Symbol
LSM14A
Entrez Gene ID
26065
Gene Name
LSM14A, mRNA processing body assembly factor
Gene Aliases
C19orf13, FAM61A, RAP55, RAP55A
Chromosome Location
Chr. 19: 34172428 - 34229515 on Build GRCh38
UniGene ID
Hs.744009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114093.1 NP_001107565.1 4 708
NM_015578.2 NP_056393.2 4 708
XM_005258719.4 XP_005258776.1 4 727
XM_005258720.4 XP_005258777.1 4 727
XM_005258721.4 XP_005258778.1 4 727
XM_011526708.2 XP_011525010.1 4 727
XM_017026575.1 XP_016882064.1 4 727
XM_017026576.1 XP_016882065.1 4 727
XM_017026577.1 XP_016882066.1 4 727
GenBank mRNA AB069974.1 - 4 587
AB069975.1 - 4 587
AK027369.1 - 3 318
AK027643.1 - 4 589
AK127401.1 - 1 831
AL833221.1 - 2 181
AL834398.1 - 4 617
BC016842.1 - 4 708 3902704

View Full Product Details