Gene Transcripts

Gene Symbol
UPF2
Entrez Gene ID
26019
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
HUPF2, RENT2, smg-3
Chromosome Location
Chr. 10: 11920022 - 12043170 on Build GRCh38
UniGene ID
Hs.370689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015542.3 NP_056357.1 19, 20 3645
NM_080599.2 NP_542166.1 19, 20 3815
XM_017016034.1 XP_016871523.1 18, 19 3301
GenBank mRNA AB037829.1 - 18, 19 4014
AF301013.1 - 19, 20 3638
AF318574.1 - 19, 20 3615
AK000764.1 - 4, 5 476
AK074375.1 - 4, 5 408
AK295209.1 - 6, 7 714
AL080198.1 - 13, 14 1815
AY013249.1 - 18, 19 3564
BC114964.1 - 19, 20 3609 40082739
BC115737.1 - 19, 20 3609 40082728

View Full Product Details