Gene Transcripts
- Gene Symbol
- TBC1D3P1-DHX40P1
- Entrez Gene ID
- 653645
- Gene Name
- TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
- Gene Aliases
- DHX40P, DHX40P1, TBC1D3P1
- Chromosome Location
- Chr. 17: 59962363 - 60019052 on Build GRCh38
- UniGene ID
- Hs.448884
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002924.3 |
|
13 |
1457 |
|
- Gene Symbol
- RNFT1
- Entrez Gene ID
- 51136
- Gene Name
- ring finger protein, transmembrane 1
- Gene Aliases
- PTD016
- Chromosome Location
- Chr. 17: 59952362 - 59964756 on Build GRCh38
- UniGene ID
- Hs.531701
- Species
- Homo sapiens
View Full Product Details