Gene Transcripts

Gene Symbol
OR4C12
Entrez Gene ID
283093
Gene Name
olfactory receptor family 4 subfamily C member 12
Gene Aliases
OR11-259
Chromosome Location
Chr. 11: 49981473 - 49982535 on Build GRCh38
UniGene ID
Hs.553669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005270.4 NP_001005270.3 1 765
GenBank mRNA BC136850.1 - 1 765
BC136851.1 - 1 765 9020842

View Full Product Details