Gene Transcripts
- Gene Symbol
- ARL17A
- Entrez Gene ID
- 51326
- Gene Name
- ADP ribosylation factor like GTPase 17A
- Gene Aliases
- ARF1P2, ARL17P1
- Chromosome Location
- Chr. 17: 46500213 - 46579781 on Build GRCh38
- UniGene ID
- Hs.559259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF208864.1 |
- |
4 |
422 |
|
View Full Product Details