Gene Transcripts

Gene Symbol
Spryd7
Entrez Gene ID
290303
Gene Name
SPRY domain containing 7
Gene Aliases
Clld6, RGD1306437
Chromosome Location
Chr. 15: 41871144 - 41890684 on Build Rnor_6.0
UniGene ID
Rn.163304
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009635.1 NP_001009635.1 4 552
GenBank mRNA BC088471.1 - 4 552

View Full Product Details