Gene Transcripts

Gene Symbol
Acbd6
Entrez Gene ID
289125
Gene Name
acyl-CoA binding domain containing 6
Gene Aliases
Chromosome Location
Chr. 13: 73196507 - 73334077 on Build Rnor_6.0
UniGene ID
Rn.27349
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011906.1 NP_001011906.1 6 1153
XM_006250031.2 XP_006250093.1 4 1020
GenBank mRNA BC086598.1 - 6 1153

View Full Product Details