Gene Transcripts

Gene Symbol
Rab35
Entrez Gene ID
288700
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 12: 46704274 - 46718405 on Build Rnor_6.0
UniGene ID
Rn.18008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013046.1 NP_001013064.1 2, 3 207
XM_008769221.1 XP_008767443.1 2, 3 257
GenBank mRNA BC085769.1 - 2, 3 207

View Full Product Details