Gene Transcripts

Gene Symbol
Bod1
Entrez Gene ID
287173
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
Fam44b, RGD1305622
Chromosome Location
Chr. 10: 16259728 - 16266069 on Build Rnor_6.0
UniGene ID
Rn.144696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013854.1 NP_001013876.2 4 1443
GenBank mRNA BC079025.1 - 4 1443
FQ213681.1 - 4 1242

View Full Product Details