Gene Transcripts

Gene Symbol
Wdr70
Entrez Gene ID
294783
Gene Name
WD repeat domain 70
Gene Aliases
RGD1309487
Chromosome Location
Chr. 2: 56944462 - 57196522 on Build Rnor_6.0
UniGene ID
Rn.17260
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013909.1 NP_001013931.1 11 1121
GenBank mRNA BC089903.1 - 11 1121

View Full Product Details