Gene Transcripts

Gene Symbol
Ugp2
Entrez Gene ID
289827
Gene Name
UDP-glucose pyrophosphorylase 2
Gene Aliases
Chromosome Location
Chr. 14: 106208174 - 106249322 on Build Rnor_6.0
UniGene ID
Rn.3415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024743.1 NP_001019914.1 5 756
XM_006251538.1 XP_006251600.1 5 708
GenBank mRNA BC097369.1 - 5 756

View Full Product Details