Gene Transcripts

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
Chromosome Location
Chr. X: 71911073 - 72153286 on Build GRCh38
UniGene ID
Hs.397836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530933.1 XP_011529235.1 6 3464
GenBank mRNA BC033261.1 - 2 3676
BC136756.1 - 3 2885 9020746

View Full Product Details