Gene Transcripts

Gene Symbol
Vegfb
Entrez Gene ID
89811
Gene Name
vascular endothelial growth factor B
Gene Aliases
Chromosome Location
Chr. 1: 222237000 - 222242742 on Build Rnor_6.0
UniGene ID
Rn.198550
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053549.1 NP_446001.1 7 920
XM_006230911.2 XP_006230973.1 7 1119
GenBank mRNA BC076395.1 - 7 920

View Full Product Details