Gene Transcripts

Gene Symbol
Mrgprb5
Entrez Gene ID
404644
Gene Name
MAS-related G protein-coupled receptor, member B5
Gene Aliases
MrgB5
Chromosome Location
Chr. 1: 103544783 - 103545844 on Build Rnor_6.0
UniGene ID
Rn.141968
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002284.1 NP_001002284.1 1 817

View Full Product Details