Gene Transcripts

Gene Symbol
Vom1r111
Entrez Gene ID
494242
Gene Name
vomeronasal 1 receptor 111
Gene Aliases
V1rf4
Chromosome Location
Chr. 1: 61219351 - 61220256 on Build Rnor_6.0
UniGene ID
Rn.129451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009522.1 NP_001009522.1 1 346
GenBank mRNA AY510366.1 - 1 346

Gene Symbol
Vom1r17
Entrez Gene ID
100312945
Gene Name
vomeronasal 1 receptor 17
Gene Aliases
Chromosome Location
Chr. 1: 60958006 - 60958745 on Build Rnor_6.0
UniGene ID
Rn.129451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167572.1 NP_001161044.1 346

View Full Product Details