Gene Transcripts

Gene Symbol
Ankrd13c
Entrez Gene ID
433667
Gene Name
ankyrin repeat domain 13c
Gene Aliases
AI505652, AU022220
Chromosome Location
Chr. 3: 157946668 - 158006837 on Build GRCm38
UniGene ID
Mm.265806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013806.1 NP_001013828.1 13 2000
GenBank mRNA AK135906.1 - 14 1736
AK157542.1 - 2182
BC065088.1 - 12 4194

View Full Product Details