Gene Transcripts

Gene Symbol
Cnrip1
Entrez Gene ID
380686
Gene Name
cannabinoid receptor interacting protein 1
Gene Aliases
1500041B16Rik, 3110054C06Rik, 5330437A18Rik, AI854501, C2orf32
Chromosome Location
Chr. 11: 17051934 - 17079372 on Build GRCm38
UniGene ID
Mm.425666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029861.2 NP_084137.1 2 534
GenBank mRNA AK005381.1 - 3 598
AK148349.1 - 2 610
AK171947.1 - 2 584
BC087946.1 - 2 535 6591410

View Full Product Details