Gene Transcripts

Gene Symbol
C8g
Entrez Gene ID
69379
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
Chromosome Location
Chr. 2: 25498650 - 25501926 on Build GRCm38
UniGene ID
Mm.24596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271777.1 NP_001258706.1 7 669
NM_027062.2 NP_081338.1 7 890
GenBank mRNA AK005962.1 - 7 673
BC019967.1 - 7 890 4023907

View Full Product Details