Gene Transcripts

Gene Symbol
Btbd17
Entrez Gene ID
72014
Gene Name
BTB (POZ) domain containing 17
Gene Aliases
1500005I02Rik, Tango10a
Chromosome Location
Chr. 11: 114790669 - 114802640 on Build GRCm38
UniGene ID
Mm.38347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028055.4 NP_082331.1 3 2005
GenBank mRNA AK032100.1 - 3 2006

View Full Product Details