Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
74441
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
4933404A18Rik, C79179
Chromosome Location
Chr. 1: 97059447 - 97128340 on Build GRCm38
UniGene ID
Mm.60362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028942.4 NP_083218.1 8 1441
GenBank mRNA AK016647.1 - 8 1389
AK029505.1 - 8 1442
AK132668.1 - 8 1392
AK161249.1 - 8 1392
BC139066.1 - 8 1344 8862088
BC139067.1 - 8 1344 8862089

View Full Product Details