Gene Transcripts

Gene Symbol
Tmco5b
Entrez Gene ID
75275
Gene Name
transmembrane and coiled-coil domains 5B
Gene Aliases
4930563P21Rik
Chromosome Location
Chr. 2: 113285095 - 113297190 on Build GRCm38
UniGene ID
Mm.159484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029232.2 NP_083508.1 11, 12 850
GenBank mRNA AK016214.1 - 11, 12 841
BC050803.1 - 11, 12 850 6771315

View Full Product Details