Gene Transcripts

Gene Symbol
Ubl5
Entrez Gene ID
66177
Gene Name
ubiquitin-like 5
Gene Aliases
1110030M22Rik, beacon
Chromosome Location
Chr. 9: 20643318 - 20646790 on Build GRCm38
UniGene ID
Mm.390165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025401.3 NP_079677.1 1 195
GenBank mRNA AK040449.1 - 1 179
AK051149.1 - 1 196
AK154036.1 - 1 637

View Full Product Details