Gene Transcripts

Gene Symbol
SEC61A2
Entrez Gene ID
55176
Gene Name
Sec61 translocon alpha 2 subunit
Gene Aliases
Chromosome Location
Chr. 10: 12129641 - 12169958 on Build GRCh38
UniGene ID
Hs.112955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142627.2 NP_001136099.1 3 248
NM_018144.3 NP_060614.2 3 248
NR_024576.2 3 248
NR_024577.2 3 248
GenBank mRNA AF346603.1 - 3 189
AK022640.1 - 3 183
AK057532.1 - 3 189
AK292295.1 - 3 198
BC026179.1 - 3 198 4823130
BP212860.1 - 3 248

View Full Product Details