Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
Chromosome Location
Chr. 5: 66596348 - 67169595 on Build GRCh38
UniGene ID
Hs.595458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164664.1 NP_001158136.1 25 3736
NM_001290226.1 NP_001277155.1 23 3188
NM_001290227.1 NP_001277156.1 22 2759
NM_001297651.1 NP_001284580.1 22 3220
NM_015183.2 NP_055998.1 24 2929
XM_006714606.3 XP_006714669.1 23 3182
XM_006714610.2 XP_006714673.1 21 2968
XM_011543382.2 XP_011541684.1 27 3828
XM_011543384.2 XP_011541686.1 22 4887
XM_011543385.2 XP_011541687.1 22 6007
XM_011543386.2 XP_011541688.1 22 2838
XM_017009447.1 XP_016864936.1 28 3909
XM_017009448.1 XP_016864937.1 26 3792
XM_017009449.1 XP_016864938.1 23 3227
XM_017009450.1 XP_016864939.1 23 2921
XM_017009451.1 XP_016864940.1 22 2720
XM_017009452.1 XP_016864941.1 23 6236
XM_017009453.1 XP_016864942.1 22 13051
GenBank mRNA AB002301.1 - 14 1972
AK296707.1 - 22 2724
AY830839.1 - 24 2930
BX648980.1 - 4 538

View Full Product Details