Gene Transcripts

Gene Symbol
TMEM240
Entrez Gene ID
339453
Gene Name
transmembrane protein 240
Gene Aliases
C1orf70, SCA21
Chromosome Location
Chr. 1: 1534778 - 1540360 on Build GRCh38
UniGene ID
Hs.668654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114748.1 NP_001108220.1 3 190
GenBank mRNA BC147018.1 - 4 219
BC147020.1 - 4 219 9056948
BU929336.1 - 3 171

View Full Product Details