Gene Transcripts

Gene Symbol
C6orf99
Entrez Gene ID
100130967
Gene Name
chromosome 6 open reading frame 99
Gene Aliases
yR211F11.1
Chromosome Location
Chr. 6: 158869865 - 158918115 on Build GRCh38
UniGene ID
Hs.32804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195032.1 NP_001181961.1 2, 3 250
NM_001302839.1 NP_001289768.1 2, 3 584
NM_001302840.1 NP_001289769.1 3, 4 345
NM_001302841.1 NP_001289770.1 2, 3 244
XM_005266788.4 XP_005266845.1 2, 3 591
XM_017010142.1 XP_016865631.1 2, 3 181
XM_017010143.1 XP_016865632.1 2, 3 258
XM_017010144.1 XP_016865633.1 2, 3 253
XM_017010145.1 XP_016865634.1 1, 2 156
XM_017010146.1 XP_016865635.1 1, 2 151
XM_017010147.1 XP_016865636.1 2, 3 241
GenBank mRNA BF688381.1 - 2, 3 251
HY052980.1 - 3, 4 346
W37337.1 - 2, 3 172

View Full Product Details