Gene Transcripts

Gene Symbol
MIS18BP1
Entrez Gene ID
55320
Gene Name
MIS18 binding protein 1
Gene Aliases
C14orf106, HSA242977, KNL2, M18BP1
Chromosome Location
Chr. 14: 45203190 - 45253402 on Build GRCh38
UniGene ID
Hs.732769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018353.4 NP_060823.3 2 713
XM_005267833.4 XP_005267890.1 3 760
XM_017021425.1 XP_016876914.1 4 741
GenBank mRNA AB067490.1 - 2 713
AK002048.1 - 2 509
BC051885.1 - 2 490
BC065544.1 - 2 504 6015439

View Full Product Details