Gene Transcripts

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
Chromosome Location
Chr. 7: 92246216 - 92401773 on Build GRCh38
UniGene ID
Hs.83293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019004.1 NP_061877.1 7, 8 1456
XM_005250458.3 XP_005250515.1 7, 8 1474
XM_011516334.1 XP_011514636.1 6, 7 1291
XM_011516335.2 XP_011514637.1 7, 8 1038
XM_017012359.1 XP_016867848.1 7, 8 1474
GenBank mRNA AB037807.1 - 7, 8 1456
AK025475.1 - 4, 5 553
BC063861.1 - 7, 8 1432
BC073893.1 - 1, 2 48

View Full Product Details