Gene Transcripts

Gene Symbol
WSB2
Entrez Gene ID
55884
Gene Name
WD repeat and SOCS box containing 2
Gene Aliases
SBA2
Chromosome Location
Chr. 12: 118032687 - 118062430 on Build GRCh38
UniGene ID
Hs.506985
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278557.1 NP_001265486.1 2, 3 245
NM_018639.4 NP_061109.1 2, 3 345
XM_017019642.1 XP_016875131.1 2, 3 247
GenBank mRNA AF163324.1 - 1, 2 182
AF229181.1 - 2, 3 284
AK294230.1 - 1, 2 103
AK295555.1 - 2, 3 246
AK310892.1 - 2, 3 223
BC015887.2 - 2, 3 235 3910968

View Full Product Details