Gene Transcripts

Gene Symbol
PYDC2
Entrez Gene ID
152138
Gene Name
pyrin domain containing 2
Gene Aliases
POP2, cPOP2
Chromosome Location
Chr. 3: 191461163 - 191461456 on Build GRCh38
UniGene ID
Hs.690618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083308.1 NP_001076777.1 1 88
GenBank mRNA AY858112.1 - 1 88

View Full Product Details