Gene Transcripts

Gene Symbol
ENDOD1
Entrez Gene ID
23052
Gene Name
endonuclease domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 95089810 - 95132651 on Build GRCh38
UniGene ID
Hs.167115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015036.2 NP_055851.1 2 2357
GenBank mRNA AB020637.1 - 2 2308

View Full Product Details