Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018920.3 | NP_061743.1 | 1 | 254 | |
NM_032087.2 | NP_114476.1 | 1 | 254 | ||
GenBank mRNA | AF152327.1 | - | 1 | 93 | |
AF152514.1 | - | 1 | 93 | ||
BC136747.1 | - | 1 | 158 | 9020737 | |
BC136748.1 | - | 1 | 158 | 9020738 | |
DA287863.1 | - | 1 | 254 |