Gene Transcripts
- Gene Symbol
- LOC100509620
- Entrez Gene ID
- 100509620
- Gene Name
- putative aquaporin-7-like protein 3
- Gene Aliases
- Chromosome Location
- Chr. 2: 94587240 - 94613324 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- AQP7
- Entrez Gene ID
- 364
- Gene Name
- aquaporin 7
- Gene Aliases
- AQP7L, AQPap, GLYCQTL
- Chromosome Location
- Chr. 9: 33383135 - 33402682 on Build GRCh38
- UniGene ID
- Hs.455323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055744.1 |
- |
1 |
1358 |
|
- Gene Symbol
- AQP7P1
- Entrez Gene ID
- 375719
- Gene Name
- aquaporin 7 pseudogene 1
- Gene Aliases
- bA251O17.3
- Chromosome Location
- Chr. 9: 63315243 - 63334520 on Build GRCh38
- UniGene ID
- Hs.658215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002817.2 |
|
4 |
1409 |
|
GenBank mRNA |
AK128231.1 |
- |
4 |
1405 |
|
BC070322.1 |
- |
4 |
732 |
|
- Gene Symbol
- AQP7P5
- Entrez Gene ID
- 102724655
- Gene Name
- aquaporin 7 pseudogene 5
- Gene Aliases
- Chromosome Location
- Chr. 9: 40463754 - 40499011 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details