Gene Transcripts
- Gene Symbol
- BRWD1
- Entrez Gene ID
- 54014
- Gene Name
- bromodomain and WD repeat domain containing 1
- Gene Aliases
- C21orf107, DCAF19, N143, WDR9
- Chromosome Location
- Chr. 21: 39184176 - 39321595 on Build GRCh38
- UniGene ID
- Hs.627139
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007246.2 |
NP_001007247.1
|
4, 5 |
495 |
|
NM_018963.4 |
NP_061836.2
|
4, 5 |
495 |
|
NM_033656.3 |
NP_387505.1
|
4, 5 |
495 |
|
XM_011529611.1 |
XP_011527913.1
|
4, 5 |
528 |
|
XM_017028373.1 |
XP_016883862.1
|
5, 6 |
300 |
|
XM_017028375.1 |
XP_016883864.1
|
4, 5 |
528 |
|
GenBank mRNA |
AB080586.1 |
- |
4, 5 |
299 |
|
AB080587.1 |
- |
4, 5 |
299 |
|
AJ238214.2 |
- |
4, 5 |
299 |
|
AJ292465.1 |
- |
4, 5 |
279 |
|
AJ292466.1 |
- |
4, 5 |
279 |
|
BC064602.1 |
- |
4, 5 |
331 |
|
BC142986.1 |
- |
4, 5 |
344 |
|
BI914338.1 |
- |
|
341 |
|
DA792365.1 |
- |
4, 5 |
531 |
|
View Full Product Details