Gene Transcripts

Gene Symbol
Ly49i3
Entrez Gene ID
494208
Gene Name
immunoreceptor Ly49i3
Gene Aliases
Chromosome Location
Chr. 4: 164743490 - 164747388 on Build Rnor_6.0
UniGene ID
Rn.55469
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009499.1 NP_001009499.1 79
GenBank mRNA AY659932.1 - 79

View Full Product Details