Gene Transcripts

Gene Symbol
Znrf4
Entrez Gene ID
301127
Gene Name
zinc and ring finger 4
Gene Aliases
Chromosome Location
Chr. 9: 10535340 - 10536528 on Build Rnor_6.0
UniGene ID
Rn.104114
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024878.1 NP_001020049.1 1183
GenBank mRNA BC098025.1 - 1183

View Full Product Details