Gene Transcripts

Gene Symbol
Rbx1
Entrez Gene ID
300084
Gene Name
ring-box 1, E3 ubiquitin protein ligase
Gene Aliases
Chromosome Location
Chr. 7: 122700849 - 122710907 on Build Rnor_6.0
UniGene ID
Rn.8068
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034135.1 NP_001029307.1 490
GenBank mRNA BC100258.1 - 490

View Full Product Details