Gene Transcripts

Gene Symbol
Tmem45b
Entrez Gene ID
315524
Gene Name
transmembrane protein 45b
Gene Aliases
RGD1305586
Chromosome Location
Chr. 8: 32563872 - 32609212 on Build Rnor_6.0
UniGene ID
Rn.42073
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033067.1 NP_001028239.1 3 560
GenBank mRNA BC100635.1 - 3 560

View Full Product Details