Gene Transcripts

Gene Symbol
Myl2
Entrez Gene ID
363925
Gene Name
myosin, light chain 2, regulatory, cardiac, slow
Gene Aliases
Mlc-2, Mlc2
Chromosome Location
Chr. 12: 39951863 - 39959065 on Build Rnor_6.0
UniGene ID
Rn.37176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035252.2 NP_001030329.2 4 253
GenBank mRNA BC126064.1 - 4 253
M11851.1 - 4 265
X07314.1 - 4 246

View Full Product Details