Gene Transcripts
- Gene Symbol
- POGLUT1
- Entrez Gene ID
- 56983
- Gene Name
- protein O-glucosyltransferase 1
- Gene Aliases
- C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
- Chromosome Location
- Chr. 3: 119468938 - 119494708 on Build GRCh38
- UniGene ID
- Hs.231750
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152305.2 |
NP_689518.1
|
1, 2 |
170 |
|
NR_024265.1 |
|
1, 2 |
170 |
|
XM_017006879.1 |
XP_016862368.1
|
1, 2 |
116 |
|
XM_017006880.1 |
XP_016862369.1
|
1, 2 |
116 |
|
GenBank mRNA |
AF168711.1 |
- |
1, 2 |
121 |
|
AK075444.1 |
- |
1, 2 |
106 |
|
AK222937.1 |
- |
1, 2 |
130 |
|
AK295984.1 |
- |
1, 2 |
145 |
|
AK301002.1 |
- |
1, 2 |
145 |
|
AK315367.1 |
- |
1, 2 |
145 |
|
AY298903.1 |
- |
1, 2 |
156 |
|
AY358581.1 |
- |
1, 2 |
113 |
|
BC030614.2 |
- |
1, 2 |
156 |
4796951 |
BC048810.1 |
- |
1, 2 |
114 |
5243487 |
BP347179.1 |
- |
1, 2 |
170 |
|
View Full Product Details