Gene Transcripts

Gene Symbol
Nicn1
Entrez Gene ID
619581
Gene Name
nicolin 1
Gene Aliases
Chromosome Location
Chr. 8: 117062905 - 117068136 on Build Rnor_6.0
UniGene ID
Rn.190831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034999.1 NP_001030171.1 6 1822
XM_006243854.2 XP_006243916.1 7 1072
GenBank mRNA BC101861.1 - 6 1822
FQ229763.1 - 1 915

View Full Product Details