Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001029929.2 | NP_001025100.1 | 6 | 1685 | |
GenBank mRNA | AK029988.1 | - | 6 | 1686 | |
AK128979.1 | - | 5 | 1992 | ||
AK171931.1 | - | 6 | 1534 | ||
BC038060.1 | - | 3 | 295 | ||
BC151086.1 | - | 6 | 1357 | 9087999 |