Gene Transcripts

Gene Symbol
OR6B3
Entrez Gene ID
150681
Gene Name
olfactory receptor family 6 subfamily B member 3
Gene Aliases
OR6B3P, OR6B3Q
Chromosome Location
Chr. 2: 240044611 - 240053722 on Build GRCh38
UniGene ID
Hs.626616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173351.1 NP_775486.1 1 824
XM_011510671.2 XP_011508973.1 3 1221

View Full Product Details