Gene Transcripts

Gene Symbol
Ly96
Entrez Gene ID
448830
Gene Name
lymphocyte antigen 96
Gene Aliases
MD-2
Chromosome Location
Chr. 5: 1972212 - 1989448 on Build Rnor_6.0
UniGene ID
Rn.141496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024279.1 NP_001019450.1 284
GenBank mRNA AY963291.1 - 284

View Full Product Details